2-Hydrazinopyrazine 98%

Lieferant: Thermo Scientific
Achtung

H64086.03 H64086.06
H64086.03EA 53.2 EUR
H64086.03 H64086.06
2-Hydrazinopyrazine 98%
2-Hydrazinopyrazine

Purity: 98% CAS No.: 54608-52-5 Molecular Formula: C4H6N4 Formula Weight: 110.12 Physical Form: Solid Appearance: Yellow Melting Point: 109-113° Beilstein Reference: 606630 MDL No.: MFCD04114555

Formel: C₄H₆N₄
Molecular Weight: 110,12 g/mol
Schmelzpunkt: 109…113 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04114555
CAS-Nummer: 54608-52-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR