3-Cyclopropylmethoxy-4-difluoromethoxybenzoic acid 98%
Lieferant: Thermo Fisher Scientific
Achtung
Synonyme: RoflumilastRelated Compound D, 3-(Cyclopropylmethoxy)-4-(difluoromethoxy)benzoic acid
H64098.03 H64098.06
H64098.03EA 74.5 EUR
H64098.03 H64098.06
3-Cyclopropylmethoxy-4-difluoromethoxybenzoic acid 98%
3-Cyclopropylmethoxy-4-difluoromethoxybenzoic acid
Purity: 98% CAS No.: 162401-62-9 Molecular Formula: C12H12F2O4 Formula Weight: 258.22 Physical Form: Solid Melting Point: 118-120° Beilstein Reference: 11413145 MDL No.: MFCD04621687
Formel: C₁₂H₁₂F₂O₄
Molecular Weight: 258.221 g/mol
Schmelzpunkt: 118…120 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04621687
CAS-Nummer: 162401-62-9
EINECS: 605-288-7