5-Brom-2-chloranilin 98%

Lieferant: Thermo Fisher Scientific
Gefahr

H64104.06 H64104.14 H64104.22
H64104.06EA 45.1 EUR
H64104.06 H64104.14 H64104.22
5-Brom-2-chloranilin 98%
5-Brom-2-chloranilin

Purity: 98% CAS No.: 60811-17-8 Molecular Formula: C6H5BrClN Formula Weight: 206.47 Physical Form: Solid Melting Point: 145-150° Beilstein Reference: 2716274 UN No.: UN2811 MDL No.: MFCD06796283

Formel: C₆H₅BrClN
Molecular Weight: 206.47 g/mol
Schmelzpunkt: 145…150 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06796283
CAS-Nummer: 60811-17-8
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR