5-Brom-2-chlor-3-methylpyridin 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5-Brom-2-chlor-3-picolin
H64109.06
H64109.14
H64109.06EA
43.8
EUR
H64109.06
H64109.14
5-Brom-2-chlor-3-methylpyridin 98%
5-Brom-2-chlor-3-methylpyridin
Purity: 98%
CAS No.: 29241-60-9
Molecular Formula: C6H5BrClN
Formula Weight: 206.47
Physical Form: Solid
Melting Point: 40-44°
Beilstein Reference: 1562796
MDL No.: MFCD03095093
Formel:
C₆H₅BrClN Molecular Weight: 206,47 g/mol Schmelzpunkt: 40…44 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03095093 CAS-Nummer: 29241-60-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...