6-Chloro-2-pyridinecarboxaldehyde 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 6-Chloropyridine-2-carboxaldehyde, 6-Chloro-2-formylpyridine, 6-Chloropicolinaldehyde

H64129.03 H64129.06
H64129.03EA 141 EUR
H64129.03 H64129.06
6-Chloro-2-pyridinecarboxaldehyde 97%
6-Chloro-2-pyridinecarboxaldehyde

Purity: 97% CAS No.: 54087-03-5 Molecular Formula: C6H4ClNO Formula Weight: 141.55 Physical Form: Powder Appearance: White Melting Point: 69-70° Beilstein Reference: 1446583 MDL No.: MFCD09832941

Formel: C₆H₄ClNO
Molecular Weight: 141,55 g/mol
Schmelzpunkt: 69…70 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09832941
CAS-Nummer: 54087-03-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR