9,9'-Spirobi[9H-fluorene] 98%

Lieferant: Thermo Scientific

Synonyme: 9,9'-Spirobi[fluorene], 9,9'-Spirobifluorene

H64133.03 H64133.06
H64133.03EA 60.2 EUR
H64133.03 H64133.06
9,9'-Spirobi[9H-fluorene] 98%
9,9'-Spirobi[9H-fluorene]

Purity: 98% CAS No.: 159-66-0 Molecular Formula: C25H16 Formula Weight: 316.40 Physical Form: Solid Appearance: White to pale yellow Melting Point: 195-200° Beilstein Reference: 1914463 MDL No.: MFCD08704216

Formel: C₂₅H₁₆
Molecular Weight: 316,40 g/mol
Schmelzpunkt: 195…200 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08704216
CAS-Nummer: 159-66-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR