4-Bromophthalimide ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5-Bromoisoindoline-1,3-dione, 5-bromo-2,3-dihydro-1H-isoindole-1,3-dione
H64136.06
H64136.14
H64136.06EA
60.7
EUR
H64136.06
H64136.14
4-Bromophthalimide ≥97%
4-Bromophthalimide
Purity: 97+%
CAS No.: 6941-75-9
Molecular Formula: C8H4BrNO2
Formula Weight: 226.03
Physical Form: Solid
Appearance: Off-white
Melting Point: 230-235°
Beilstein Reference: 131235
MDL No.: MFCD00466049
Formel:
C₈H₄BrNO₂ Molecular Weight: 226,03 g/mol Schmelzpunkt: 230…235 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00466049 CAS-Nummer: 6941-75-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...