2-Cyan-6-methylpyridin 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
6-Methyl-2-pyridincarbonitril, 6-Cyan-2-picolin
H64148.06
H64148.14
H64148.06EA
57.5
EUR
H64148.06
H64148.14
2-Cyan-6-methylpyridin 98%
2-Cyano-6-methylpyridine
Purity: 98%
CAS No.: 1620-75-3
Molecular Formula: C7H6N2
Formula Weight: 118.14
Physical Form: Solid
Appearance: Off-white
Melting Point: 70-74°
Beilstein Reference: 111107
UN No.: UN3439
MDL No.: MFCD00190586
Formel:
C₇H₆N₂ Molecular Weight: 118.14 g/mol Schmelzpunkt: 70…74 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00190586 CAS-Nummer: 1620-75-3 EINECS: 216-588-9 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...