2,3,5-Trichlorpyridin 98%

Lieferant: Thermo Scientific

Synonyme: 2,3,5-TCP

H64173.22 H64173.36
H64173.22EA 80.8 EUR
H64173.22 H64173.36
2,3,5-Trichlorpyridin 98%
2,3,5-Trichlorpyridin

Purity: 98+% CAS No.: 16063-70-0 Molecular Formula: C5H2Cl3N Formula Weight: 182.43 Storage Temperature: 2°C to 8°C Flash Point: 113°(235°F) Physical Form: Solid Appearance: Off-white to pale yellow Melting Point: 46-50° lit. Boiling Point: 219° lit. Beilstein Reference: 119384 MDL No.: MFCD00043007

Formel: C₅H₂Cl₃N
Molecular Weight: 182,44 g/mol
Siedepunkt: 219 °C (Lit.)
Schmelzpunkt: 46…50 °C (Lit.)
Flammpunkt: 113 °C (235 °F)
Lagertemperatur: Kühlschrank
MDL: MFCD00043007
CAS-Nummer: 16063-70-0
EINECS: 407-270-2
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR