4-Formylbenzeneboronic acid pinacol ester 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Formylphenylboronic acid pinacol ester, 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde, 4-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde

H64192.03 H64192.06 H64192.14
H64192.03EA 38.9 EUR
H64192.03 H64192.06 H64192.14
4-Formylbenzeneboronic acid pinacol ester 98%
4-Formylbenzeneboronic acid pinacol ester

Purity: 98% CAS No.: 128376-64-7 Molecular Formula: C13H17BO3 Formula Weight: 232.09 Physical Form: Crystalline solid Appearance: Off-white Melting Point: 54-59° Beilstein Reference: 7704448 MDL No.: MFCD04972375

Formel: C₁₃H₁₇BO₃
Molecular Weight: 232,086 g/mol
Schmelzpunkt: 54…59 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04972375
CAS-Nummer: 128376-64-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR