2-Fluor-5-nitropyridin 98%
Lieferant: Thermo Scientific
|
Achtung
|
H64210.06
H64210.14
H64210.06EA
53.4
EUR
H64210.06
H64210.14
2-Fluor-5-nitropyridin 98%
2-Fluoro-5-nitropyridine
Purity: 98%
CAS No.: 456-24-6
Molecular Formula: C5H3FN2O2
Formula Weight: 142.09
Physical Form: Liquid
Density: 1.41
Refractive Index: 1.5250
Sensitivity: Moisture Sensitive
Appearance: Colourless to pale yellow
Melting Point: 17-21°
Boiling Point: 86-87°/7mm lit.
Beilstein Reference: 120351
MDL No.: MFCD03095059
Formel:
C₅H₃FN₂O₂ Siedepunkt: 86…87 °C (7 mmHg) (Lit.) Schmelzpunkt: 17…21 °C Dichte: 1,41 Lagertemperatur: Raumtemperatur |
MDL:
MFCD03095059 CAS-Nummer: 456-24-6 EINECS: 207-260-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...