3-Brom-9-phenyl-9H-carbazol 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-Bromo-9-phenylcarbazole

H64212.06 H64212.14 H64212.22
H64212.06EA 49.5 EUR
H64212.06 H64212.14 H64212.22
3-Brom-9-phenyl-9H-carbazol 98%
3-Bromo-9-phenyl-9H-carbazole

Purity: 98% CAS No.: 1153-85-1 Molecular Formula: C18H12BrN Formula Weight: 322.20 Physical Form: Solid Melting Point: 97-98° Beilstein Reference: 1315492 MDL No.: MFCD11977305

Formel: C18H12BrN
Molecular Weight: 322.20 g/mol
Schmelzpunkt: 97…98 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11977305
CAS-Nummer: 1153-85-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR