2-Chloro-1-nitro-3,5-bis(trifluoromethyl)benzene 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3,5-Bis(trifluoromethyl)-2-chloronitrobenzene
H64221.03
H64221.06
H64221.03EA
50.6
EUR
H64221.03
H64221.06
2-Chloro-1-nitro-3,5-bis(trifluoromethyl)benzene 98%
2-Chloro-1-nitro-3,5-bis(trifluoromethyl)benzene
Purity: 98%
CAS No.: 654-55-7
Molecular Formula: C8H2ClF6NO2
Formula Weight: 293.55
Physical Form: Solid
Beilstein Reference: 2888862
MDL No.: MFCD04972677
Formel:
C₈H₂ClF₆NO₂ Molecular Weight: 293,549 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04972677 CAS-Nummer: 654-55-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...