4-Azaindol 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1H-Pyrrolo[2,3-e]pyridin
H64224.03
H64224.06
H64224.03EA
70.5
EUR
H64224.03
H64224.06
4-Azaindol 97%
4-Azaindol
Purity: 97%
CAS No.: 272-49-1
Molecular Formula: C7H6N2
Formula Weight: 118.14
Physical Form: Solid
Melting Point: 125-127°
Beilstein Reference: 109683
MDL No.: MFCD00971977
Formel:
C₇H₆N₂ Schmelzpunkt: 125…127 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00971977 CAS-Nummer: 272-49-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...