4-Brompicolinsäure 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Bromopyridine-2-carboxylic acid

H64244.03 H64244.06 H64244.14
H64244.03EA 39.1 EUR
H64244.03 H64244.06 H64244.14
4-Brompicolinsäure 97%
4-Brompicolinsäure

Purity: 97% CAS No.: 30766-03-1 Molecular Formula: C6H4BrNO2 Formula Weight: 202.01 Melting Point: 177-179° Beilstein Reference: 471891 MDL No.: MFCD03426665

Formel: C₆H₄BrNO₂
Molecular Weight: 202,01 g/mol
Schmelzpunkt: 177…179 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03426665
CAS-Nummer: 30766-03-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR