2,4-Dichlor-6-methylpyridin 97+%

Lieferant: Thermo Fisher Scientific
Achtung

H64248.03 H64248.06
H64248.03EA 83.9 EUR
H64248.03 H64248.06
2,4-Dichlor-6-methylpyridin 97+%
2,4-Dichloro-6-methylpyridine

Purity: 97+% CAS No.: 42779-56-6 Molecular Formula: C6H5Cl2N Formula Weight: 162.01 Storage Temperature: 2°C to 8°C Boiling Point: 205-206° Beilstein Reference: 111084 MDL No.: MFCD08669834

Formel: C₆H₅Cl₂N
Molecular Weight: 162.01 g/mol
Siedepunkt: 205…206 °C
MDL: MFCD08669834
CAS-Nummer: 42779-56-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR