Methyl-3-(4-hydroxyphenyl)propanoat 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-(4-Hydroxyphenyl)propionic Acid Methyl Ester, Methyl 3-(4-hydroxyphenyl)propanoate, Methyl 3-(4-hydroxyphenyl)propionate, 3-(4-Hydroxy-phenyl)-propionic acid-OMe
H64255.14
H64255.22
H64255.14EA
102
EUR
H64255.14
H64255.22
Methyl-3-(4-hydroxyphenyl)propanoat 98%
Methyl-3-(4-hydroxyphenyl)propionate
Purity: 98+%
CAS No.: 5597-50-2
Molecular Formula: C10H12O3
Formula Weight: 180.20
Flash Point:
Formel:
C₁₀H₁₂O₃ Molecular Weight: 180,20 g/mol Siedepunkt: 108 °C (11 mmHg) Schmelzpunkt: 39…41 °C (Lit.) Flammpunkt: >113 °C (235 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00071577 CAS-Nummer: 5597-50-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...