3-Brompicolinsäure 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-Bromopyridine-2-carboxylic acid, 3-Bromo-2-pyridinecarboxylic acid
H64258.06
H64258.14
H64258.06EA
88.9
EUR
H64258.06
H64258.14
3-Brompicolinsäure 97%
3-Brompicolinsäure
Purity: 97%
CAS No.: 30683-23-9
Molecular Formula: C6H4BrNO2
Formula Weight: 202.01
Physical Form: Solid
Density: 1.813
Melting Point: 141-144°
Beilstein Reference: 472171
MDL No.: MFCD01320380
Formel:
C₆H₄BrNO₂ Molecular Weight: 202,01 g/mol Schmelzpunkt: 141…144 °C Dichte: 1,813 Lagertemperatur: Raumtemperatur |
MDL:
MFCD01320380 CAS-Nummer: 30683-23-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...