5-Methyluridin 99%
Lieferant: Thermo Fisher Scientific
Purity: 99%
CAS No.: 1463-10-1
Molecular Formula: C10H14N2O6
Formula Weight: 258.23
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: Off-white to white
Melting Point: 183-184°
Beilstein Reference: 90164
Optical Rotation: -8°(C=2 in water)
MDL No.: MFCD00006535
Formel:
C₁₀H₁₄N₂O₆ Schmelzpunkt: 183…184 °C |
MDL:
MFCD00006535 CAS-Nummer: 1463-10-1 EINECS: 215-973-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...