5-Azaindol 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1H-pyrrolo[3,2-c]pyridine, 1H-Pyrrolo[2,3-d]pyridine
H64272.03
H64272.06
H64272.03EA
47
EUR
H64272.03
H64272.06
5-Azaindol 98%
5-Azaindol
Purity: 98%
CAS No.: 271-34-1
Molecular Formula: C7H6N2
Formula Weight: 118.14
Melting Point: 104-108°
Beilstein Reference: 109684
MDL No.: MFCD00955936
Formel:
C₇H₆N₂ Molecular Weight: 118,14 g/mol Schmelzpunkt: 104…108 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00955936 CAS-Nummer: 271-34-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...