6-fluoronicotinonitrile 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5-Cyano-2-fluoropyridine, 6-Fluoronicotinonitrile
H64278.03
H64278.06
H64278.03EA
74.5
EUR
H64278.03
H64278.06
6-fluoronicotinonitrile 95%
6-fluoronicotinonitrile
Purity: 95%
CAS No.: 3939-12-6
Molecular Formula: C6H3FN2
Formula Weight: 122.10
Physical Form: Solid
Appearance: Off-white
Melting Point: 51-52°
Beilstein Reference: 471855
UN No.: UN3439
MDL No.: MFCD09261087
Formel:
C₆H₃FN₂ Molecular Weight: 122,102 g/mol Schmelzpunkt: 51…52 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09261087 CAS-Nummer: 3939-12-6 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...