Ethyl-3-amino-3-iminopropanoate hydrochloride 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: Ethyl 2-amidinoacetate hydrochloride

H64288.06 H64288.14
H64288.06EA 104 EUR
H64288.06 H64288.14
Ethyl-3-amino-3-iminopropanoate hydrochloride 95%
Ethyl-3-amino-3-iminopropanoate hydrochloride

Purity: 95% CAS No.: 57508-48-2 Molecular Formula: C5H10N2O2.HCl Formula Weight: 166.60 Physical Form: Solid Melting Point: 105-108° Beilstein Reference: 3694030 MDL No.: MFCD06797615

Formel: C₅H₁₁ClN₂O₂
Molecular Weight: 166,60 g/mol
Schmelzpunkt: 105…108 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06797615
CAS-Nummer: 57508-48-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR