(2S)-2-Oxiranylmethyl-3-nitrobenzolsulfonat 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: (S)-(+)-Glycidyl nosylate, (S)-Oxiran-2-ylmethyl 3-nitrobenzenesulphonate, Glycidyl (S)-(+)-3-nitrobenzenesulphonate

H64289.06 H64289.14
H64289.06EA 90 EUR
H64289.06 H64289.14
(2S)-2-Oxiranylmethyl-3-nitrobenzolsulfonat 98%
(S)-Oxiran-2-ylmethyl-3-nitrobenzenesulphonate

Purity: 98% CAS No.: 115314-14-2 Molecular Formula: C9H9NO6S Formula Weight: 259.23 Melting Point: 63-66° Beilstein Reference: 4704457 MDL No.: MFCD00064582

Formel: C₉H₉NO₆S
Molecular Weight: 259,23 g/mol
Schmelzpunkt: 63…66 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064582
CAS-Nummer: 115314-14-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR