3-Brom-4-methoxypyridin 97%
Lieferant: Thermo Scientific
|
Achtung
|
H64296.03
H64296.06
H64296.03EA
127
EUR
H64296.03
H64296.06
3-Brom-4-methoxypyridin 97%
3-Bromo-4-methoxypyridine
Purity: 97%
CAS No.: 82257-09-8
Molecular Formula: C6H6BrNO
Formula Weight: 188.02
Storage Temperature: -30°C to -10°C
Physical Form: Liquid
Appearance: Pale yellow
Beilstein Reference: 1423610
MDL No.: MFCD05664024
Formel:
C₆H₆BrNO |
MDL:
MFCD05664024 CAS-Nummer: 82257-09-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...