Methyl-4-brompicolinat 97+%

Lieferant: Thermo Scientific
Achtung

Synonyme: Methyl 4-bromopicolinate, Methyl 4-bromopyridine-2-carboxylate

H64327.03 H64327.06 H64327.14
H64327.03EA 42.8 EUR
H64327.03 H64327.06 H64327.14
Methyl-4-brompicolinat 97+%
Methyl-4-brompicolinat

Purity: 97+% CAS No.: 29681-42-3 Molecular Formula: C7H6BrNO2 Formula Weight: 216.03 Storage Temperature: 2°C to 8°C Physical Form: Solid Melting Point: 54-56° Beilstein Reference: 473047 MDL No.: MFCD07374897

Formel: C₇H₆BrNO₂
Molecular Weight: 216,03 g/mol
Schmelzpunkt: 54…56 °C
MDL: MFCD07374897
CAS-Nummer: 29681-42-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR