3-Brom-6-chlor-2-methylpyridin 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Brom-2-chlor-6-picolin, 5-Brom-2-chlor-6-methylpyridin, 3-Brom-6-chlor-2-picolin

H64333.06 H64333.14
H64333.06EA 25.9 EUR
H64333.06 H64333.14
3-Brom-6-chlor-2-methylpyridin 98%
3-Bromo-6-chloro-2-methylpyridine

Purity: 98% CAS No.: 132606-40-7 Molecular Formula: C6H5BrClN Formula Weight: 206.47 Physical Form: Liquid Melting Point: 46-47° Boiling Point: 80-84°/2mm lit. Beilstein Reference: 4243997 MDL No.: MFCD07375117

Formel: C₆H₅BrClN
Molecular Weight: 206,47 g/mol
Siedepunkt: 80…84 °C (2 mmHg) (Lit.)
Schmelzpunkt: 46…47 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07375117
CAS-Nummer: 132606-40-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR