Benzo[b]furan-3-boronsäure 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Benzofuran-3-ylboronic acid
H64389.MD
H64389.03
H64389.06
H64389.MDEA
92
EUR
H64389.MD
H64389.03
H64389.06
Benzo[b]furan-3-boronsäure 98%
Benzo[b]furan-3-boronsäure
Purity: 98%
CAS No.: 317830-83-4
Molecular Formula: C8H7BO3
Formula Weight: 161.95
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Beilstein Reference: 9479377
MDL No.: MFCD06801687
Formel:
C₈H₇BO₃ Molecular Weight: 161,95 g/mol |
MDL:
MFCD06801687 CAS-Nummer: 317830-83-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...