Methyl-6-bromo-5-fluoronicotinate 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Methyl 6-bromo-5-fluoronicotinate
H64395.03
H64395.06
H64395.03EA
98.8
EUR
H64395.03
H64395.06
Methyl-6-bromo-5-fluoronicotinate 98%
Methyl-6-bromo-5-fluoronicotinate
Purity: 98%
CAS No.: 1214336-88-5
Molecular Formula: C7H5BrFNO2
Formula Weight: 234.02
Storage Temperature: -30°C to -10°C
MDL No.: MFCD14698110
Formel:
C₇H₅BrFNO₂ |
MDL:
MFCD14698110 CAS-Nummer: 1214336-88-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...