3,6-Dihydrothiopyran-4-boronic acid pinacol ester 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3,6-Dihydro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-thiopyran, 2-(3,6-Dihydro-2H-thiopyran-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
H64416.MD
H64416.03
H64416.06
H64416.MDEA
59.7
EUR
H64416.MD
H64416.03
H64416.06
3,6-Dihydrothiopyran-4-boronic acid pinacol ester 98%
3,6-Dihydrothiopyran-4-boronic acid pinacol ester
Purity: 98%
CAS No.: 862129-81-5
Molecular Formula: C11H19BO2S
Formula Weight: 226.14
Physical Form: Solid
Beilstein Reference: 11090000
MDL No.: MFCD10700154
Formel:
C₁₁H₁₉BO₂S Molecular Weight: 226,141 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10700154 CAS-Nummer: 862129-81-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...