1-Aminocyclobutanecarboxylic acid 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-Aminocyclobutane-1-carboxylic acid, 1-Amino-1-cyclobutanecarboxylic acid
H64459.03
H64459.06
H64459.03EA
53.8
EUR
H64459.03
H64459.06
1-Aminocyclobutanecarboxylic acid 95%
1-Aminocyclobutanecarboxylic acid
Purity: 95%
CAS No.: 22264-50-2
Molecular Formula: C5H9NO2
Formula Weight: 115.13
Physical Form: Powder
Appearance: White
Melting Point: 261° dec.
Beilstein Reference: 2802253
MDL No.: MFCD00661068
Formel:
C₅H₉NO₂ Molecular Weight: 115,132 g/mol Schmelzpunkt: 261 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00661068 CAS-Nummer: 22264-50-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...