3-Methoxyphenethylamin 97+%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2-(3-Methoxyphenyl)ethylamine
H64494.06
H64494.14
H64494.22
H64494.06EA
41.6
EUR
H64494.06
H64494.14
H64494.22
3-Methoxyphenethylamin 97+%
3-Methoxyphenethylamin
Purity: 97+%
CAS No.: 2039-67-0
Molecular Formula: C9H13NO
Formula Weight: 151.21
Flash Point:
Formel:
C₉H₁₃NO Molecular Weight: 151.21 g/mol Siedepunkt: 118…118 °C (6 mmHg) Dichte: 1.038 Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008187 CAS-Nummer: 2039-67-0 EINECS: 218-017-9 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...