Isopropyl benzenesulphonate 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: Benzenesulphonic acid isopropyl ester

H64523.03 H64523.06 H64523.14
H64523.03EA 53.3 EUR
H64523.03 H64523.06 H64523.14
Isopropyl benzenesulphonate 95%
Isopropyl benzenesulphonate

Purity: 95% CAS No.: 6214-18-2 Molecular Formula: C9H12O3S Formula Weight: 200.25 Storage Temperature: 2°C to 8°C Physical Form: Liquid Refractive Index: 1.5020 Appearance: Colourless to pale yellow Beilstein Reference: 1106314 MDL No.: MFCD04038771

Formel: C₉H₁₂O₃S
Molecular Weight: 200.252 g/mol
MDL: MFCD04038771
CAS-Nummer: 6214-18-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR