9,9'-Spirobi[9H-fluorene]-2-boronic acid 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 9,9'-Spirobifluorene-2-boronic acid

H64525.03 H64525.06
H64525.03EA 93.2 EUR
H64525.03 H64525.06
9,9'-Spirobi[9H-fluorene]-2-boronic acid 98%
9,9'-Spirobi[9H-fluorene]-2-boronic acid

Purity: 98% CAS No.: 236389-21-2 Molecular Formula: C25H17BO2 Formula Weight: 360.22 Physical Form: Powder Appearance: Off-white Beilstein Reference: 10478712 MDL No.: MFCD13248568

Formel: C₂₅H₁₇BO₂
Molecular Weight: 360,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13248568
CAS-Nummer: 236389-21-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR