3-oxobutanenitrile 96%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Oxobutanenitrile

H64536.MD H64536.03
H64536.MDEA 57.8 EUR
H64536.MD H64536.03
3-oxobutanenitrile 96%
3-oxobutanenitrile

Purity: 96% CAS No.: 2469-99-0 Molecular Formula: C4H5NO Formula Weight: 83.09 Physical Form: Liquid Refractive Index: 1.4285 Appearance: Colourless to pale yellow Boiling Point: 92-94°/10mm Beilstein Reference: 1633584 UN No.: UN3276 MDL No.: MFCD05861381

Formel: C₄H₅NO
Molecular Weight: 83,09 g/mol
Siedepunkt: 92…94 °C (10 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD05861381
CAS-Nummer: 2469-99-0
EINECS: 219-590-8
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR