2-Brom-6-chlorbenzaldehyd 98%

Lieferant: Thermo Scientific
Achtung

H64605.03 H64605.06
H64605.03EA 105 EUR
H64605.03 H64605.06
2-Brom-6-chlorbenzaldehyd 98%
2-Bromo-6-chlorobenzaldehyde

Purity: 98% CAS No.: 64622-16-8 Molecular Formula: C7H4BrClO Formula Weight: 219.46 Physical Form: Solid Melting Point: 81-86° Beilstein Reference: 2613087 MDL No.: MFCD00266792

Formel: C₇H₄BrClO
Molecular Weight: 219,46 g/mol
Schmelzpunkt: 81…86 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00266792
CAS-Nummer: 64622-16-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR