3-Brombenzamid 99%

Lieferant: Thermo Fisher Scientific
Achtung

H64672.14 H64672.22
H64672.14EA 57.8 EUR
H64672.14 H64672.22
3-Brombenzamid 99%
3-Brombenzamid

Purity: 99% CAS No.: 22726-00-7 Molecular Formula: C7H6BrNO Formula Weight: 200.04 Physical Form: Solid Appearance: White Melting Point: 156-159° Beilstein Reference: 1860187 MDL No.: MFCD00017127

Formel: C₇H₆BrNO
Molecular Weight: 200.04 g/mol
Schmelzpunkt: 156…159 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017127
CAS-Nummer: 22726-00-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR