5-Brom-2-hydroxy-4-picolin 97%
Lieferant: Thermo Scientific
Achtung
Synonyme: 5-Bromo-2-hydroxy-4-methylpyridine
H64674.06 H64674.14
H64674.06EA 54 EUR
H64674.06 H64674.14
5-Brom-2-hydroxy-4-picolin 97%
5-Brom-2-hydroxy-4-picolin
Purity: 97% CAS No.: 164513-38-6 Molecular Formula: C6H6BrNO Formula Weight: 188.02 Physical Form: Solid Appearance: Off-white Melting Point: 198-202° Beilstein Reference: 6798531 MDL No.: MFCD03427664
Formel: C₆H₆BrNO
Molecular Weight: 188,02 g/mol
Schmelzpunkt: 198…202 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03427664
CAS-Nummer: 164513-38-6