2-Brom-6-fluorbenzaldehyd 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H64689.06
H64689.14
H64689.22
H64689.06EA
49.9
EUR
H64689.06
H64689.14
H64689.22
2-Brom-6-fluorbenzaldehyd 98%
2-Bromo-6-fluorobenzaldehyde
Purity: 98%
CAS No.: 360575-28-6
Molecular Formula: C7H4BrFO
Formula Weight: 203.01
Storage Temperature: 2°C to 8°C
Flash Point:
Formel:
C₇H₄BrFO Molecular Weight: 203.01 g/mol Siedepunkt: 100…102 °C (8 mmHg) Schmelzpunkt: 43…47 °C Dichte: 1.70 Flammpunkt: >110 °C (230 °F) |
MDL:
MFCD03407341 CAS-Nummer: 360575-28-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...