2,2,2,3'-Tetrafluoracetophenon 98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2,2,2-Trifluoro-1-(3-fluorophenyl)ethanone

H64704.03 H64704.06
H64704.03EA 36.6 EUR
H64704.03 H64704.06
2,2,2,3'-Tetrafluoracetophenon 98%
2,2,2,3'-Tetrafluoracetophenon

Purity: 98% CAS No.: 708-64-5 Molecular Formula: C8H4F4O Formula Weight: 192.11 Storage Temperature: 2°C to 8°C Flash Point: 17°(63°F) Physical Form: Liquid Density: 1.372 Refractive Index: 1.4440 Appearance: Colourless to pale yellow Boiling Point: 79°/50mm Beilstein Reference: 1872788 UN No.: UN1224 MDL No.: MFCD00039236

Formel: C₈H₄F₄O
Molecular Weight: 192,11 g/mol
Siedepunkt: 79 °C (50 mmHg)
Dichte: 1,372
Flammpunkt: 17 °C (63 °F)
MDL: MFCD00039236
CAS-Nummer: 708-64-5
EINECS: 211-903-6
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR