tert-Butyl hydrogen succinate 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Succinic acid mono-tert-butyl ester, 4-(tert-Butoxy)-4-oxobutanoic acid, tert-Butyl Hydrogen Succinate, Mono-tert-butyl Succinate

H64706.03 H64706.06
H64706.03EA 40.6 EUR
H64706.03 H64706.06
tert-Butyl hydrogen succinate 97%
tert-Butyl hydrogen succinate

Purity: 97% CAS No.: 15026-17-2 Molecular Formula: C8H14O4 Formula Weight: 174.20 Flash Point: 110°(230°F) Physical Form: Solid Appearance: Yellow-brown Melting Point: 51-54° lit. Beilstein Reference: 1772169 MDL No.: MFCD00273441

Formel: C₈H₁₄O₄
Molecular Weight: 174,196 g/mol
Schmelzpunkt: 51…54 °C (Lit.)
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00273441
CAS-Nummer: 15026-17-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR