2-Brom-9,9-dimethyl-9H-fluoren 98%
Lieferant: Thermo Scientific
|
Achtung
|
H64732.06
H64732.14
H64732.22
H64732.06EA
56.7
EUR
H64732.06
H64732.14
H64732.22
2-Brom-9,9-dimethyl-9H-fluoren 98%
2-Bromo-9,9-dimethylfluorene
Purity: 98%
CAS No.: 28320-31-2
Molecular Formula: C15H13Br
Formula Weight: 273.17
Physical Form: Solid
Melting Point: 57-62°
Beilstein Reference: 2524870
MDL No.: MFCD09261272
Formel:
C₁₅H₁₃Br Molecular Weight: 273,17 g/mol Schmelzpunkt: 57…62 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09261272 CAS-Nummer: 28320-31-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...