6-Chloro-5-fluoropicolinic acid 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Chloro-3-fluoropyridine-6-carboxylic acid

H64734.03 H64734.06
H64734.03EA 53.1 EUR
H64734.03 H64734.06
6-Chloro-5-fluoropicolinic acid 98%
6-Chloro-5-fluoropicolinic acid

Purity: 98% CAS No.: 860296-24-8 Molecular Formula: C6H3ClFNO2 Formula Weight: 175.54 Physical Form: Solid Appearance: Off-white Beilstein Reference: 10063182 MDL No.: MFCD13185819

Formel: C₆H₃ClFNO₂
Molecular Weight: 175.543 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13185819
CAS-Nummer: 860296-24-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR