1-Ethyl-3-methylimidazoliumdiethylphosphat ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 1-ethyl-3-methylimidazolium diethylphosphate

H27400.18 H27400.06
H27400.18EA 411 EUR
H27400.18 H27400.06
1-Ethyl-3-methylimidazoliumdiethylphosphat ≥98%
1-Ethyl-3-methylimidazolium diethylphosphate
Formel: C₁₀H₂₁N₂O₄P
Molecular Weight: 264,26 g/mol
Schmelzpunkt: 19…21 °C
Dichte: 1,157
Lagertemperatur: Raumtemperatur
MDL: MFCD09953486
CAS-Nummer: 848641-69-0
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR