4,5,6,7-Tetrahydro-3H-imidazo[4,5-c]pyridine hydrochloride 95%
Lieferant: Thermo Fisher Scientific
Achtung
Synonyme: 4,5,6,7-Tetrahydro-5-azabenzimidazole hydrochloride
H64755.MD H64755.03
H64755.MDEA 107 EUR
H64755.MD H64755.03
4,5,6,7-Tetrahydro-3H-imidazo[4,5-c]pyridine hydrochloride 95%
4,5,6,7-Tetrahydro-3H-imidazo[4,5-c]pyridine hydrochloride
Purity: 95% CAS No.: 879668-17-4 Molecular Formula: C6H9N3.HCl Formula Weight: 159.62 Physical Form: Solid Beilstein Reference: 15312806 MDL No.: MFCD11846200
Formel: C₆H₁₀ClN₃
Molecular Weight: 159.62 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11846200
CAS-Nummer: 879668-17-4