3-Fluorphenethylamin 97+%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2-(3-Fluorophenyl)ethylamine
H64762.06
H64762.14
H64762.06EA
42.1
EUR
H64762.06
H64762.14
3-Fluorphenethylamin 97+%
3-Fluorphenethylamin
Purity: 97+%
CAS No.: 404-70-6
Molecular Formula: C8H10FN
Formula Weight: 139.17
Flash Point: 83°(181°F)
Physical Form: Liquid
Density: 1.066
Refractive Index: 1.5090
Boiling Point: 87°/15mm
Beilstein Reference: 3045789
UN No.: UN2735
MDL No.: MFCD00075376
Formel:
C₈H₁₀FN Molecular Weight: 139.17 g/mol Siedepunkt: 87 °C (15 mmHg) Dichte: 1.066 Flammpunkt: 83 °C (181 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00075376 CAS-Nummer: 404-70-6 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...