Ethyl-4-formyl-1H-pyrrole-2-carboxylate 96%

Lieferant: Thermo Scientific
Achtung

Synonyme: Ethyl 4-formylpyrrole-2-carboxylate, Ethyl 4-formyl-1H-pyrrole-2-carboxylate

H64776.03 H64776.06
H64776.03EA 61.7 EUR
H64776.03 H64776.06
Ethyl-4-formyl-1H-pyrrole-2-carboxylate 96%
Ethyl-4-formyl-1H-pyrrole-2-carboxylate

Purity: 96% CAS No.: 7126-57-0 Molecular Formula: C8H9NO3 Formula Weight: 167.16 Melting Point: 101-105° Beilstein Reference: 473842 MDL No.: MFCD04122505

Formel: C₈H₉NO₃
Molecular Weight: 167,164 g/mol
Schmelzpunkt: 101…105 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04122505
CAS-Nummer: 7126-57-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR