2-(Chlormethyl)benzothiazol 95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2-(Chloromethyl)-1,3-benzothiazole
H64781.03
H64781.06
H64781.03EA
132
EUR
H64781.03
H64781.06
2-(Chlormethyl)benzothiazol 95%
2-(Chloromethyl)benzothiazole
Purity: 95%
CAS No.: 37859-43-1
Molecular Formula: C8H6ClNS
Formula Weight: 183.65
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Sensitivity: Moisture Sensitive
Melting Point: 34°
Boiling Point: 88-99°/0.3mm
Beilstein Reference: 120805
MDL No.: MFCD05664944
Formel:
C₈H₆CINS Molecular Weight: 183.65 g/mol Siedepunkt: 88…99 °C (0.3 mmHg) |
MDL:
MFCD05664944 CAS-Nummer: 37859-43-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...