1,2-Dichlor-4-fluor-5-nitrobenzol 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4,5-Dichloro-2-fluoronitrobenzene

H64782.06 H64782.14 H64782.22
H64782.06EA 46.6 EUR
H64782.06 H64782.14 H64782.22
1,2-Dichlor-4-fluor-5-nitrobenzol 95%
1,2-Dichloro-4-fluoro-5-nitrobenzene

Purity: 95% CAS No.: 2339-78-8 Molecular Formula: C6H2Cl2FNO2 Formula Weight: 209.99 Flash Point: 113°(235°F) Physical Form: Low melting solid Density: 1.596 Refractive Index: 1.5750 Appearance: Pale yellow Melting Point: 17° Boiling Point: 247° Beilstein Reference: 3269124 MDL No.: MFCD00075330

Formel: C₆H₂Cl₂FNO₂
Molecular Weight: 209.99 g/mol
Siedepunkt: 247 °C
Dichte: 1.596
Flammpunkt: >113 °C (235 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00075330
CAS-Nummer: 2339-78-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR