Pyrazine-2,5-dicarboxylic acid 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2,5-Pyrazinedicarboxylic acid

H64795.03 H64795.06
H64795.03EA 82.3 EUR
H64795.03 H64795.06
Pyrazine-2,5-dicarboxylic acid 95%
Pyrazine-2,5-dicarboxylic acid

Purity: 95% CAS No.: 122-05-4 Molecular Formula: C6H4N2O4 Formula Weight: 168.11 Physical Form: Solid Melting Point: 255-260° lit. Beilstein Reference: 139985 MDL No.: MFCD01630902

Formel: C₆H₄N₂O₄
Molecular Weight: 168,108 g/mol
Schmelzpunkt: 255…260 °C (Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD01630902
CAS-Nummer: 122-05-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR