1-Bromo-5-fluoropentane 99%

Lieferant: Thermo Scientific
Achtung

H64806.03 H64806.06
H64806.03EA 96 EUR
H64806.03 H64806.06
1-Bromo-5-fluoropentane 99%
1-Bromo-5-fluoropentane

Purity: 99% CAS No.: 407-97-6 Molecular Formula: C5H10BrF Formula Weight: 169.04 Physical Form: Liquid Density: 1.360 Refractive Index: 1.4406 Appearance: Colourless Boiling Point: 162° Beilstein Reference: 1697258 MDL No.: MFCD01709395

Formel: C₅H₁₀BrF
Siedepunkt: 162 °C
Dichte: 1,360
Lagertemperatur: Raumtemperatur
MDL: MFCD01709395
CAS-Nummer: 407-97-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR